Order Entry
Austria
ContactUsLinkComponent
 
2,4,5-T-butyl ester (2,4,5-trichlorophenoxy acetic acid butyl ester)
Catalog # EHERC17105500
Supplier:  EHRENSTORFER
CAS Number:  
2,4,5-T-butyl ester (2,4,5-trichlorophenoxy acetic acid butyl ester)
Catalog # EHERC17105500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :311.59 g/mol
EINECS:202-277-5
CAS Number:93-79-8
Formula:C₁₂H₁₃Cl₃O₃
MDL Number:MFCD00128016