Order Entry
Austria
ContactUsLinkComponent
 
Pesticide reference standard, Monocrotophos (Dimethyl-1-methyl-2-(methylcarbamoyl)vinyl phosphate) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERXA15300000AL
 :  
Pesticide reference standard, Monocrotophos (Dimethyl-1-methyl-2-(methylcarbamoyl)vinyl phosphate) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERXA15300000AL
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    6923-22-4
  • Description:
    Monocrotophos (Dimethyl-1-methyl-2-(methylcarbamoyl)vinyl phosphate) 100 µg/ml in acetonitrile
  • Formula:
    C₇H₁₄NO₅P
  • Description:
    Monocrotophos (Dimethyl-1-methyl-2-(methylcarbamoyl)vinyl phosphate)