Cacodylic acid (dimethylarsinic acid), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-60-5
:
Formula:(CH₃)₂As(O)OH
MDL Number:MFCD00002095
:138 g/mol
Storage Temperature:Ambient
EINECS:200-883-4
UN:1572
ADR:6.1,II
:GHS08
:GHS07
Frequently Bought Together