Cacodylic acid (dimethylarsinic acid) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-60-5
:
Formula:(CH₃)₂As(O)OH
MDL Number:MFCD00002095
:138 g/mol
Melting Pt:195…198 °C
Storage Temperature:Ambient
EINECS:200-883-4
UN:1572
ADR:6.1,II
:GHS06
:GHS09
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:195 to 198 °C
- Titration with NaOH:≥97.5 %
Specification Test Results
Frequently Bought Together