Pesticide reference standard, Dieldrin 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL12590000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:60-57-1
- Description:Dieldrin 10 µg/ml in cyclohexane
- Formula:C₁₂H₈Cl₆O
- Description:Dieldrin