Order Entry
Austria
ContactUsLinkComponent
 
Specpure® Hafnium Standard Solution, 10000 mg/L Hf in 5% Hydrochloric Acid (from HfOCl₂), Standard for ICP
Catalog # 14374.AE
CAS Number:  
Specpure® Hafnium Standard Solution, 10000 mg/L Hf in 5% Hydrochloric Acid (from HfOCl₂), Standard for ICP
Catalog # 14374.AE
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    5% HCl
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00011032
  • UN No.:
    1789
  • Storage temperature:
    Ambient
  • Physical form:
    Liquid
  • Molecular formula:
    HfCl₂O
  • Element:
    Hafnium
  • Symbol:
    Hf

Specifications

About this item

Certifications: Traceable to NIST.