Order Entry
Austria
ContactUsLinkComponent
Hafnium standard solution, 1,000 mg/L Hf in 5% hydrofluoric acid standard for AAS
Hafnium standard solution, 1,000 mg/L Hf in 5% hydrofluoric acid standard for AAS
Catalog # ACRO196011000
CAS Number:  
Hafnium standard solution, 1,000 mg/L Hf in 5% hydrofluoric acid standard for AAS
Catalog # ACRO196011000
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Hafnium
  • Symbol:
    Hf

Specifications

About this item