Order Entry
Austria
ContactUsLinkComponent
 
Sodium standard solution, 1000 mg/L Na in water (from NaCl), standard for ion chromatography, Specpure®
Catalog # 41696.AE
CAS Number:  
Sodium standard solution, 1000 mg/L Na in water (from NaCl), standard for ion chromatography, Specpure®
Catalog # 41696.AE
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00044631
  • Element:
    Sodium
  • Symbol:
    Na

Specifications

About this item

Molecular Formula: NaCl in H₂O
Physical Form: Liquid
MDL No.: MFCD00044631

Certifications: Traceable to NIST