Order Entry
Austria
ContactUsLinkComponent
 
Molybdenum standard solution, 10 mg/L Mo in 5% nitric acid with hydrofluoric acid (max. 1%) (from Mo), standard for ICP, Specpure®
  45260.AE
 :  
Molybdenum standard solution, 10 mg/L Mo in 5% nitric acid with hydrofluoric acid (max. 1%) (from Mo), standard for ICP, Specpure®
  45260.AE
 :  

 

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃/tr HF
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Molybdenum
  • Symbol:
    Mo

 

 

 : Traceable to NIST