Order Entry
Austria
ContactUsLinkComponent
Purpurin(1,2,4-trihydroxy-9,10-anthraquinone) 90% (dye content), Sigma-Aldrich®
Irritant
Purpurin(1,2,4-trihydroxy-9,10-anthraquinone) 90% (dye content), Sigma-Aldrich®
Catalog # 229148-5G
CAS Number:  
undefined
Irritant
Purpurin(1,2,4-trihydroxy-9,10-anthraquinone) 90% (dye content), Sigma-Aldrich®
Catalog # 229148-5G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance (Colour):
    Conforms to Requirements
    Dark Red to Orange-Brown or Brown
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength:
    254 - 260 nm
  • Extinction Coefficient:
    ≥17000
  • Carbon:
    58.4 - 72.8%
  • Purity (HPLC):
    Typically 70 - 85%
  • Solubility (Turbidity):
    Clear to Turbid
    c = 1 mg/ml, NH₄OH
  • Solubility (Colour):
    Conforms to Requirements
    Red to Very Dark Red to Purple

Specification Test Results

About this item

 :
Melting Pt:253 - 256 °C (lit.)
Flash Pt:113 °C (closed cup)
 :256.21 g/mol
EINECS:201-359-8
 :GHS07
Storage Temperature:Room Temperature
CAS Number:81-54-9
Formula:C₁₄H₈O₅
MDL Number:MFCD00001203
Melting Pt:253…256 °C (lit.)