Order Entry
Austria
ContactUsLinkComponent
5,6,7,8-Tetrahydro-2-naphthol 98%
5,6,7,8-Tetrahydro-2-naphthol 98%
Certificates
undefined
5,6,7,8-Tetrahydro-2-naphthol 98%

Specification Test Results

  • Appearance:
    Beige Crystalline mass
  • Infrared spectrum:
    Conforms
  • Melting point:
    58 to 62 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together