Herbicide reference standard, Desethyl atrazine 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA10331000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:6190-65-4
- Description:Desethyl atrazine 100 µg/ml in acetonitrile
- Formula:C₆H₁₀ClN₅
- Description:Desethyl atrazine