Order Entry
Austria
ContactUsLinkComponent
Herbicide reference standard, Dinoterb (2-tert-Butyl-4,6-dinitrophenol) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Herbicide reference standard, Dinoterb (2-tert-Butyl-4,6-dinitrophenol) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL12830000AL
Supplier:  EHRENSTORFER
CAS Number:  
Herbicide reference standard, Dinoterb (2-tert-Butyl-4,6-dinitrophenol) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL12830000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    1420-07-1
  • Description:
    Dinoterb (2-tert-Butyl-4,6-dinitrophenol) 10 µg/ml in acetonitrile
  • Formula:
    C₁₀H₁₂N₂O₅
  • Description:
    Dinoterb (2-tert-Butyl-4,6-dinitrophenol)

Specifications

About this item