Order Entry
Austria
ContactUsLinkComponent
 
Tin standard solution, 100 mg/L Sn in dil. nitric acid with hydrofluoric acid (max. 1%) standard for ICP-MS
  ACCUICPMS63N001X1
 :  
Tin standard solution, 100 mg/L Sn in dil. nitric acid with hydrofluoric acid (max. 1%) standard for ICP-MS
  ACCUICPMS63N001X1
 :  

 

  • Concentration:
    100 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Tin
  • Symbol:
    Sn