Order Entry
Austria
ContactUsLinkComponent
Selenium standard solution, 10 mg/L Se in 5% nitric acid (from Se), standard for ICP, Specpure®
Selenium standard solution, 10 mg/L Se in 5% nitric acid (from Se), standard for ICP, Specpure®
Catalog # 45263.AE
CAS Number:  
Selenium standard solution, 10 mg/L Se in 5% nitric acid (from Se), standard for ICP, Specpure®
Catalog # 45263.AE
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Environmentally Preferable:
  • Element:
    Selenium
  • Symbol:
    Se

Specifications

About this item

Certifications: Traceable to NIST