Rubidium standard solution, 1000 mg/L Rb in 5% nitric acid (from Rb₂CO₃), standard for AAS, Specpure®
Molecular Formula: Rb₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134055
: Traceable to NIST
- MDL No.:MFCD00134055
Frequently Bought Together