Neodymium standard solution, 1000 mg/L Nd in 5% nitric acid (from Nd₂O₃), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Nd₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011130
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00011130
Specifications
Related Information
Frequently Bought Together