Order Entry
Austria
ContactUsLinkComponent
Lanthanum standard solution, 1000 mg/L La in 5% nitric acid (from La), standard for AAS, Specpure®
Lanthanum standard solution, 1000 mg/L La in 5% nitric acid (from La), standard for AAS, Specpure®
Catalog # 88074.AE
CAS Number:  
Lanthanum standard solution, 1000 mg/L La in 5% nitric acid (from La), standard for AAS, Specpure®
Catalog # 88074.AE
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00011066
  • Element:
    Lanthanum
  • Symbol:
    La

Specifications

About this item

Molecular Formula: La in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011066

Certifications: Traceable to NIST