Order Entry
Austria
ContactUsLinkComponent
Iridium standard solution, 1,000 mg/L Ir in 5% hydrochloric acid standard for AAS
Iridium standard solution, 1,000 mg/L Ir in 5% hydrochloric acid standard for AAS
Catalog # ACRO196041000
CAS Number:  
Iridium standard solution, 1,000 mg/L Ir in 5% hydrochloric acid standard for AAS
Catalog # ACRO196041000
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Iridium
  • Symbol:
    Ir

Specifications

About this item