Order Entry
Austria
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Atrazine-Desethyl
Supelco® PESTANAL® Organic Reference Standard, Atrazine-Desethyl
Catalog # 36629-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Atrazine-Desethyl
Catalog # 36629-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    6190-65-4
  • Description:
    Atrazine-desethyl
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 + H332 - H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00078646
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-Amino-4-chloro-6-isopropylamino-1,3,5-triazine; Desethylatrazine
  • Molecular weight:
    187,63
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P261 - P264 - P271 - P301 + P312 - P304 + P340 + P312 - P305 + P351 + P338