Order Entry
Austria
ContactUsLinkComponent
Tungsten standard solution, mg/L W in water with ammonia (max. 1%) standard for ICP-MS
Tungsten standard solution, mg/L W in water with ammonia (max. 1%) standard for ICP-MS
Catalog # ACCUICPMS65W0.1X1
Supplier:  AccuStandard
CAS Number:  
Tungsten standard solution, mg/L W in water with ammonia (max. 1%) standard for ICP-MS
Catalog # ACCUICPMS65W0.1X1
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O/tr HF
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Tungsten
  • Symbol:
    W

Specifications

About this item