Strontium-Standardlösung, 1000 mg/L Sr in 5%iger Salpetersäure (aus Sr(NO3)2) Specpure® Standard für die ICP
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Sr(NO₃)₂) in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134060
Certifications: Rückführbar auf NIST
Specifications
- MDL No.:MFCD00134060
Specifications
Related Information
Frequently Bought Together