Samarium standard solution, 1000 mg/L Sm in 5% nitric acid (from Sm₂O₃), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Sm in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011233
Certifications: Traceable to NIST
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together