Order Entry
Austria
ContactUsLinkComponent
 
Ruthenium standard solution, 1000 mg/L Ru in 20% hydrochloric acid (from RuCl₃), standard for AAS, Specpure®
Catalog # 89117.AE
CAS Number:  
Ruthenium standard solution, 1000 mg/L Ru in 20% hydrochloric acid (from RuCl₃), standard for AAS, Specpure®
Catalog # 89117.AE
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    20% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00011207
  • Element:
    Ruthenium
  • Symbol:
    Ru

Specifications

About this item

Molecular Formula: RuCl₃ in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011207

Certifications: Traceable to NIST