Praseodymium standard solution, 10000 mg/L Pr in 5% nitric acid (from Pr₆O₁₁), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Pr₆O₁₁ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011174
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00011174
Specifications
Related Information
Frequently Bought Together