Niobium standard solution, 1000 mg/L Nb in water (from (NH₄)NbF₆) Certipur® standard for ICP, Supelco®
Artikelnr. 1.70337.0100
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- M-Clarity quality level:MQ100
- Element:Niobium
- Symbol:Nb
Specifications
About this item
Certifications: Traceable to SRM from NIST