Indium standard solution, 10 mg/L In in 5% nitric acid (from In), standard for ICP, Specpure®
Catalog # 45256.AE
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:5% HNO₃
- Pack type:Glass bottle
- Pk:100 mL
- MDL No.:MFCD00134048
- Element:Indium
- Symbol:In
Specifications
About this item
Molecular Formula: In in 2% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134048
Certifications: Traceable to NIST