Cobalt standard solution, 1000 mg/L Co in 5% nitric acid (from Co), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Co in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010935
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00010935
Specifications
Frequently Bought Together