Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid 50% (v/v) in aqueous solution
Corrosive
Irritant
Hydrochloric acid 50% (v/v) in aqueous solution
Catalog # 35607.K2
CAS Number:  
Corrosive
Irritant
Hydrochloric acid 50% (v/v) in aqueous solution
Catalog # 35607.K2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
ADR:8,II
 :36.46 g/mol
EINECS:231-595-7
UN:1789
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324