Order Entry
Austria
ContactUsLinkComponent
 
Sulphuric acid 1.25 mol/l (2.5 N) in aqueous solution, AVS TITRINORM
Catalog # 84521.320
Supplier:  VWR Collection
CAS Number:  
Sulphuric acid 1.25 mol/l (2.5 N) in aqueous solution, AVS TITRINORM
Catalog # 84521.320
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS07
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589