Silver standard solution, 1000 mg/L Ag in 5% nitric acid (from Ag), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Ag in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00003397
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00003397
Specifications
Related Information
Frequently Bought Together