Order Entry
Austria
ContactUsLinkComponent
Silver standard solution, 10 mg/L Ag in 5% nitric acid (from Ag), standard for ICP, Specpure®
Silver standard solution, 10 mg/L Ag in 5% nitric acid (from Ag), standard for ICP, Specpure®
Catalog # 45264.AE
CAS Number:  
Silver standard solution, 10 mg/L Ag in 5% nitric acid (from Ag), standard for ICP, Specpure®
Catalog # 45264.AE
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00003397
  • Element:
    Silver
  • Symbol:
    Ag

Specifications

About this item

Molecular Formula: Ag in 2% HNO₃
Physical Form: Liquid
Density: ca 1.0
UN No.: 3264
MDL No.: MFCD00003397

Certifications: Traceable to NIST