Dysprosium standard solution, 1000 mg/L Dy in 5% nitric acid (from Dy₂O₃), standard for AAS, Specpure®
Molecular Formula: Dy₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010982
: Traceable to NIST
- MDL No.:MFCD00010982
Frequently Bought Together