Order Entry
Austria
ContactUsLinkComponent
Glycerine, anhydrous ≥99.5%, Ultrapure Bioreagent for biotechnology, for electrophoresis, J.T.Baker®
Glycerine, anhydrous ≥99.5%, Ultrapure Bioreagent for biotechnology, for electrophoresis, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Glycerine, anhydrous ≥99.5%, Ultrapure Bioreagent for biotechnology, for electrophoresis, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • Assay (HOCH₂CHOHCH₂OH):
    ≥99.5 %
  • Absorbance of a 1M Solution - 280 nm:
    ≤0.050
  • DNase Activity (None Detected):
    Passes Test
  • RNase Activity (None Detected):
    Passes Test
  • Protease Activity (None Detected):
    Passes Test
  • Absorbance of a 1M Solution - Color (APHA):
    ≤10
  • Residue after Ignition:
    ≤0.005 %
  • Fatty Acid Esters (as Butyric Acid):
    ≤0.05 %
  • Trace Impurities - Iron (Fe):
    ≤5 ppm
  • Trace Impurities - Lead (Pb):
    ≤2.000 ppm
  • Trace Impurities - Mercury (Hg):
    ≤10.000 ppm
  • Trace Impurities - Zinc (Zn):
    ≤2.000 ppm

Specification Test Results

Frequently Bought Together