Order Entry
Austria
ContactUsLinkComponent
Acetonitrile ≥99.5% (by GC), AR® ACS, Macron Fine Chemicals™
Flammable
Irritant
Acetonitrile ≥99.5% (by GC), AR® ACS, Macron Fine Chemicals™
Catalog # C0043-26
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
Acetonitrile ≥99.5% (by GC), AR® ACS, Macron Fine Chemicals™
Catalog # C0043-26
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    350 lb
  • Seq:
    7251
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥99.5%
  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • ACS - Assay (CH₃CN) (by GC, corrected for water):
    ≥99.5 %
  • ACS - Appearance (clear, colorless liquid, free of extraneous matter):
    Passes Test
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤0.005 %
  • ACS - Titrable Acid (µeq/g):
    ≤8
  • ACS - Titrable Base (µeq/g):
    ≤0.6
  • ACS - Water (H₂O)(by Karl Fischer titrn):
    ≤0.3 %

Specification Test Results

About this item

 :
 :41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Density:0.79 g/cm³
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
 :GHS02
 :GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878