Order Entry
Austria
ContactUsLinkComponent
Sodium nitroprusside dihydrate ≥99% ACS, Fluka™
Toxic
Sodium nitroprusside dihydrate ≥99% ACS, Fluka™
Catalog # 228710-500G
CAS Number:  
Toxic
Sodium nitroprusside dihydrate ≥99% ACS, Fluka™
Catalog # 228710-500G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    99.0 - 102.0 %
  • Water insoluble matter:
    Max. 0.01 %
  • Chloride (Cl):
    Max. 200 ppm
  • Sulfate (SO₄):
    Max. 0.01 %

Specification Test Results

About this item

 :
Merck Index:13,08722
Density:1.72 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3288
ADR:6.1,III
 :297.95 g/mol
EINECS:238-373-9
 :GHS06
CAS Number:13755-38-9
Formula:Na₂[Fe(CN)₅NO]·2H₂O
MDL Number:MFCD00149192