Order Entry
Austria
ContactUsLinkComponent
Thallium standard solution, 1000 mg/L Tl in dil. nitric acid standard for ICP-MS
Thallium standard solution, 1000 mg/L Tl in dil. nitric acid standard for ICP-MS
Catalog # ACCUICPMS60N01X1
Supplier:  AccuStandard
CAS Number:  
Thallium standard solution, 1000 mg/L Tl in dil. nitric acid standard for ICP-MS
Catalog # ACCUICPMS60N01X1
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Thallium
  • Symbol:
    Tl

Specifications

About this item