Tellurium standard solution, 1000 mg/L Te in 20% hydrochloric acid standard for AAS
Catalog # ACCUAA58H1
Supplier: AccuStandard
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:20% HCl
- Pack type:Glass bottle
- Pk:100 mL
- Environmentally Preferable:
- Element:Tellurium
- Symbol:Te