Order Entry
Austria
ContactUsLinkComponent
 
Silicon standard solution, 1,000 mg/L Si in 2% sodium hydroxide solution standard for AAS
Catalog # ACRO196291000
CAS Number:  
Silicon standard solution, 1,000 mg/L Si in 2% sodium hydroxide solution standard for AAS
Catalog # ACRO196291000
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% NaOH
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Silicon
  • Symbol:
    Si

Specifications

About this item