Order Entry
Austria
ContactUsLinkComponent
Scandium standard solution, 1000 mg/L Sc in 5% nitric acid (from Sc₂O₃), standard for AAS, Specpure®
Scandium standard solution, 1000 mg/L Sc in 5% nitric acid (from Sc₂O₃), standard for AAS, Specpure®
Catalog # 88093.AP
CAS Number:  
Scandium standard solution, 1000 mg/L Sc in 5% nitric acid (from Sc₂O₃), standard for AAS, Specpure®
Catalog # 88093.AP
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • MDL No.:
    MFCD00016323
  • Element:
    Scandium
  • Symbol:
    Sc

Specifications

About this item

Molecular Formula: Sc₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00016323

Certifications: Traceable to NIST