Order Entry
Austria
ContactUsLinkComponent
Phosphorus standard solution, 1,000 mg/L P in water standard for ICP-MS
Phosphorus standard solution, 1,000 mg/L P in water standard for ICP-MS
Catalog # ACCUICPMS41W0.1X1
Supplier:  AccuStandard
CAS Number:  
Phosphorus standard solution, 1,000 mg/L P in water standard for ICP-MS
Catalog # ACCUICPMS41W0.1X1
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2-5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Phosphorus
  • Symbol:
    P

Specifications

About this item