Rhodium standard solution, 1000 mg/L Rh in 20% hydrochloric acid (from RhCl₃), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: RhCl₃ in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011201
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00011201
Specifications
Related Information
Frequently Bought Together