Order Entry
Austria
ContactUsLinkComponent
Mercury standard solution, 1,000 mg/l Hg in dil. nitric acid standard for AAS
Mercury standard solution, 1,000 mg/l Hg in dil. nitric acid standard for AAS
Catalog # ACCUAA34N5
Supplier:  AccuStandard
CAS Number:  
Mercury standard solution, 1,000 mg/l Hg in dil. nitric acid standard for AAS
Catalog # ACCUAA34N5
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item