Order Entry
Austria
ContactUsLinkComponent
Praseodymium standard solution, 1000 mg/L Pr in 5% nitric acid (from Pr₆O₁₁), standard for ICP, Specpure®
Praseodymium standard solution, 1000 mg/L Pr in 5% nitric acid (from Pr₆O₁₁), standard for ICP, Specpure®
Artikelnr. 13812.AE
CAS Number:  
Praseodymium standard solution, 1000 mg/L Pr in 5% nitric acid (from Pr₆O₁₁), standard for ICP, Specpure®
Artikelnr. 13812.AE
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00011174
  • Element:
    Praseodymium
  • Symbol:
    Pr

Specifications

About this item

Molecular Formula: Pr6O11 in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011174