Order Entry
Austria
ContactUsLinkComponent
Germanium standard solution, 1000 mg/L Ge in water (from (NH?)?GeF?) Certipur® standard for ICP, Supelco®
Germanium standard solution, 1000 mg/L Ge in water (from (NH?)?GeF?) Certipur® standard for ICP, Supelco®
Catalog # 1.70320.0100
Supplier:  Merck
CAS Number:  
undefined
Germanium standard solution, 1000 mg/L Ge in water (from (NH?)?GeF?) Certipur® standard for ICP, Supelco®
Catalog # 1.70320.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Germanium
  • Symbol:
    Ge

Specifications

About this item

Certifications: Traceable to SRM from NIST