ContactUsLinkComponent
Sulphur standard solution, 1000 mg/L S in water (from (NH₄)₂SO₄), standard for ICP, Specpure®
Sulphur standard solution, 1000 mg/L S in water (from (NH₄)₂SO₄), standard for ICP, Specpure®
Certificates
Sulphur standard solution, 1000 mg/L S in water (from (NH₄)₂SO₄), standard for ICP, Specpure®
Certificates
Specifications
  • MDL No.:
    MFCD00085316