Order Entry
Austria
ContactUsLinkComponent
Sulphur standard solution, 1000 mg/L S in water standard for ICP
Sulphur standard solution, 1000 mg/L S in water standard for ICP
Catalog # ACCUICP56W5
Supplier:  AccuStandard
CAS Number:  
Sulphur standard solution, 1000 mg/L S in water standard for ICP
Catalog # ACCUICP56W5
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Sulphur
  • Symbol:
    S

Specifications

About this item