Order Entry
Austria
ContactUsLinkComponent
Magnesium standard solution, 1000 mg/L Mg in 0,5 M nitric acid standard for AAS
Magnesium standard solution, 1000 mg/L Mg in 0,5 M nitric acid standard for AAS
Catalog # ACRO196105000
CAS Number:  
Magnesium standard solution, 1000 mg/L Mg in 0,5 M nitric acid standard for AAS
Catalog # ACRO196105000
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Magnesium
  • Symbol:
    Mg

Specifications

About this item