Order Entry
Austria
ContactUsLinkComponent
Magnesium sulphate, anhydrous, Reagent Grade
Magnesium sulphate, anhydrous, Reagent Grade
Catalog # 0338-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Magnesium sulphate, anhydrous, Reagent Grade
Catalog # 0338-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Moisture:
    <0.2%
  • Purity:
    >98%

Specification Test Results

About this item

Synonyms: MgSO4, Magnesium sulphate anhydrous
 :
 :120.37 g/mol
Merck Index:12,05731
Melting Pt:1127 °C
Density:2.65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-298-2
CAS Number:7487-88-9
Formula:MgSO₄
MDL Number:MFCD00011110