Order Entry
Austria
ContactUsLinkComponent
Cadmium standard solution, 1000 mg/L Cd in dil. nitric acid (from Cd) TraceCERT® standard for ICP, Supelco®
Cadmium standard solution, 1000 mg/L Cd in dil. nitric acid (from Cd) TraceCERT® standard for ICP, Supelco®
Catalog # SUPL36379-100ML-F
Supplier:  Merck
CAS Number:  
Cadmium standard solution, 1000 mg/L Cd in dil. nitric acid (from Cd) TraceCERT® standard for ICP, Supelco®
Catalog # SUPL36379-100ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Cadmium
  • Symbol:
    Cd

Specifications

About this item

Certifications: Traceable to SRM from NIST, tested in ISO 17034 /ISO 17025 accredited laboratory.